• USA Home
  • P6161 - C-Phycocyanin from Spirulina sp.

EMAIL THIS PAGE TO A FRIEND
P6161 Sigma

C-Phycocyanin from Spirulina sp.

lyophilized powder

Purchase

Properties

Related Categories Auxiliary Proteins, Biochemicals and Reagents, Phycobiliproteins, Proteins and Derivatives
form   lyophilized powder
composition   Protein, ~30%
storage temp.   2-8°C

Description

Packaging

Package size based on protein content

Physical form

Highly purified, lyophilized powder containing sucrose, dithioerythritol and sodium azide as preservatives

Biochem/physiol Actions

C-phycocyanin was shown to function as an antioxidant and to inhibit the production of melanin in human skin by reducing expression and activity of tyrosinase.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 1687 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?