• USA Home
  • P6179 - DL-Norephedrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
P6179 Sigma

DL-Norephedrine hydrochloride

Sigma Reference Standard

Synonym: (±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride

Purchase

Properties

Related Categories Alkaloid, Cell Biology, Nutrition Research, Phytochemicals by Chemical Classification
grade   Sigma Reference Standard
InChI Key   DYWNLSQWJMTVGJ-PRCZDLBKSA-N
packaging   vial of 250 mg
mp   194-196 °C(lit.)

Description

Biochem/physiol Actions

Mixed-acting sympathomimetic amine.

Packaging

Supplied in amber screw-cap vials

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
DN4200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P7876 DL-Norephedrine hydrochloride, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?