• USA Home
  • P6238 - trans-5-Phenyl-4-pentenyl hydroperoxide

EMAIL THIS PAGE TO A FRIEND
P6238 Sigma

trans-5-Phenyl-4-pentenyl hydroperoxide

≥90%, Solution in ethanol

Synonym: (E)-5-Phenyl-4-pentenyl hydroperoxide, PHPP solution

Purchase

Description

Biochem/physiol Actions

Hydroperoxide compound used to assay peroxidase activity. Reduced to 5-phenyl-4E-pentenol (PPA) by plant and animal peroxidases in the presence of reducing substrates by a reaction that can be monitored spectrophotometrically.

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
2
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?