EMAIL THIS PAGE TO A FRIEND
P6306 Sigma

V8 Protease

Purchase

Properties

sterility   aseptically filled
storage temp.   −20°C

Description

Preparation Note

Prepared from P 2922

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37°C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P2922 Endoproteinase Glu-C from Staphylococcus aureus V8, Type XVII-B, lyophilized powder, 500-1,000 units/mg solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?