EMAIL THIS PAGE TO A FRIEND
P6362 Sigma

β-Pompilidotoxin

≥98%

Synonym: β-PMTX

Purchase

Description

Amino Acid Sequence

Arg-Ile-Lys-Ile-Gly-Leu-Phe-Asp-Gln-Leu-Ser-Arg-Leu-NH2

Biochem/physiol Actions

Voltage-gated sodium channel activator.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?