• USA Home
  • P6363 - Monoclonal Anti-PMCA ATPase antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
P6363 Sigma

Monoclonal Anti-PMCA ATPase antibody produced in mouse

clone 5F10, ascites fluid

Synonym: Anti-Plasma Membrane Ca2+ ATPase

Properties

Related Categories Alphabetical Index, Antibodies, PJ-PN, Primary Antibodies
species reactivity   human, primate, canine, rat, hamster, rabbit, eel
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  immunoprecipitation: suitable
  western blot: suitable
clone   5F10, monoclonal
antibody form   ascites fluid
mol wt   antigen mol wt 140 kDa
contains   0.05% sodium azide
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... ATP2B1(490)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

human erythrocyte Ca2+ ATPase.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?