• USA Home
  • P6409 - Phosphatase, Acid, Prostatic from bovine prostate

EMAIL THIS PAGE TO A FRIEND
P6409 Sigma

Phosphatase, Acid, Prostatic from bovine prostate

lyophilized powder, ~10 units/g solid

Purchase

Description

Analysis Note

Protein determined by biuret

Other Notes

Note: 60 Sigma units = 1 μmolar unit.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 4.8 at 37 °C. Prostatic acid phosphatase activity is the difference between the total acid phosphatase activity and the acid phosphatase activity in the presence of 20 mM tartrate.

Physical form

Partially purified, lyophilized powder

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?