• USA Home
  • P6410 - Pancreatic Polypeptide rat

EMAIL THIS PAGE TO A FRIEND
P6410 Sigma

Pancreatic Polypeptide rat

≥95% (HPLC)

Synonym: Ala-Pro-Leu-Glu-Pro-Met-Tyr-Pro-Gly-Asp-Tyr-Ala-Thr-His-Glu-Gln-Arg-Ala-Gln-Tyr-Glu-Thr-Gln-Leu-Arg-Arg-Tyr-Ile-Asn-Thr-Leu-Thr-Arg-Pro-Arg-Tyr-NH2

  • CAS Number 90419-12-8

  • Empirical Formula (Hill Notation) C195H298N58O57S

  • Molecular Weight 4398.87

  •  MDL number MFCD00081813

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Gastrointestinal Peptides, Natriuretic Peptides, Pancreatic Polypeptides,
assay   ≥95% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Ppy(24677)

Description

Amino Acid Sequence

Ala-Pro-Leu-Glu-Pro-Met-Tyr-Pro-Gly-Asp-Tyr-Ala-Thr-His-Glu-Gln-Arg-Ala-Gln-Tyr-Glu-Thr-Gln-Leu-Arg-Arg-Tyr-Ile-Asn-Thr-Leu-Thr-Arg-Pro-Arg-Tyr-NH2

Biochem/physiol Actions

Agonist at Y4 neuropeptide Y receptors.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?