EMAIL THIS PAGE TO A FRIEND
P6426 Sigma

PDX G.F.

50-150 μm (dry), fractionation range 1500-30,000 Da (globular proteins)

Purchase

Properties

Related Categories Gel Filtration Chromatography, Gel Filtration Media, Molecular Biology, Protein Chromatography, Proteomics More...
swelling   1 g swells to 9-11 mL
particle size   50-150 μm (dry)
pore size   1500-30,000 Da fractionation range (globular proteins)

Description

Application

For Gel Filtration Chromatography

PDX is used in protein chromatography, gel filtration chromatograpy and gel filtration media. PDX has been used to study diabetes treatment and age-related decline of glucose tolerance.

Other Notes

Cross-linked dextran beads

Legal Information

PDX is a trademark of Polydex Biologicals

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?