• USA Home
  • P6427 - pBR322 (Hind III) Sequencing Primer, counterclockwise, 16-mer

EMAIL THIS PAGE TO A FRIEND
P6427 Sigma

pBR322 (Hind III) Sequencing Primer, counterclockwise, 16-mer

Synonym: 5′-GCA ATT TAA CTG TGA T-3′

Purchase

Description

Application

Used to sequence DNA that has been cloned into pBR322. Both clockwise and counterclockwise primers are listed for BamH I and Pst I. Because of the proximity of the EcoR I and Hind III sites, the Hind III counterclockwise primer can be used to sequence DNA that has been cloned into the EcoR I site, and the EcoR I clockwise primer can be used to sequence DNA that has been cloned into the Hind III site.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?