• USA Home
  • P6481 - Petri dishes, polystyrene
EMAIL THIS PAGE TO A FRIEND
P6481 Sigma
Petri dishes, polystyrene
size 100 mm × 15 mm, compartmentalized with vertical stacking rings
Synonym: petri dish, petri plate, petri plates
Purchase
Properties
Related Categories Cell Culture Supplies, Culture Dishes, Excel Scientific, Excel Scientific Petri Dishes, Labware,
sterility   sterile; γ-irradiated
  sterile
feature   disposable
  Optically clear with consistently flat bottoms
size   100 mm × 15 mm
Description
General description
Designed for easy one hand opening and closing, edges of tops and bottoms are squared off to provide a firm sure grip. Flare and length of lid skirt allows lid to be removed and replaced easily. Dishes have three venting ribs on the underside of the lid to prevent condensation build up.
• Standard slippable petri dish
• Side Arrows and ISO Mark Target for automation
• Disposable (single use) polystyrene petri dish for general microbiological use
• Molded from medical-grade virgin polystyrene
• Optically clear
• Radiation sterilized
• Contents sterile if sleeve is unopened
• Ventilation ribs allow for free air circulation and reduce condensation during incubation
• Flared lid skirt and squared corners for easy one-hand operation, especially with gloves
Please note: Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner; this results in a size difference in the outer diameter of approx 10 mm.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?