EMAIL THIS PAGE TO A FRIEND
P6514 Sigma

Phenylacetylene

~98%

Synonym: Ethynylbenzene

Purchase

Properties

vapor pressure   17.6 mmHg ( 37.7 °C)
InChI Key   UEXCJVNBTNXOEH-UHFFFAOYSA-N
assay   ~98%
refractive index   n20/D 1.549(lit.)
bp   142-144 °C(lit.)
density   0.93 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3295 3/PG 3
WGK Germany 
1
RTECS 
DA0780000
Flash Point(F) 
81 °F
Flash Point(C) 
27 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?