• USA Home
  • P6638 - L-α-Phosphatidylcholine

EMAIL THIS PAGE TO A FRIEND
P6638 Sigma

L-α-Phosphatidylcholine

10 mg/mL in chloroform, ~99%, from bovine brain

Synonym: 1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Glycerophosphocholines, Glycerophospholipids,
biological source   from bovine brain
assay   ~99%
concentration   10 mg/mL in chloroform
storage temp.   −20°C

Description

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.

Preparation Note

Prepared chromatographically

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
UN 1888 6.1/PG 3
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?