EMAIL THIS PAGE TO A FRIEND
P6657 Sigma

Prostaglandin D2

cell culture tested

Synonym: (5Z,9α,13E,15S)-9,15-Dihydroxy-11-oxoprosta-5,13-dien-1-oic-acid, PGD2

  • CAS Number 41598-07-6

  • Empirical Formula (Hill Notation) C20H32O5

  • Molecular Weight 352.47

  •  Beilstein Registry Number 2170623

  •  MDL number MFCD00077857

  •  PubChem Substance ID 329820251

Purchase

Properties

InChI Key   BHMBVRSPMRCCGG-OUTUXVNYSA-N
Quality Level   GMP
sterility   γ-irradiated
suitability   cell culture tested
storage temp.   −20°C

Description

Biochem/physiol Actions

Primary prostaglandin in brain; induces inflammation; stimulates adenylyl cyclase.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
UK7930000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P5172 Prostaglandin D2, ≥95%, synthetic

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?