• USA Home
  • P6895 - Tree/shrub pollens from Betula fontinalis occidentalis (water birch)

EMAIL THIS PAGE TO A FRIEND
P6895 Sigma

Tree/shrub pollens from Betula fontinalis occidentalis (water birch)

Purchase

Properties

Related Categories Cell Biology, Nutrition Research, Phytochemicals by Chemical Classification, Plant Extracts
storage temp.   2-8°C

Description

Application

Pollens, particulates that contain the microgametophytes of seed plants which produce the male gametes (sperm cells), are frequently involved in human allergy studies. Tree/shrub pollens from Betula fontinalis occidentalis (water birch) are used together with other pollens to assess human allergic response to various pollens and to assess the immunological differences among birch tree pollens.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?