• USA Home
  • P6932 - Carestream® Kodak® Processing chemicals

EMAIL THIS PAGE TO A FRIEND
P6932 Sigma

Carestream® Kodak® Processing chemicals

T-MAX developer

Purchase

Description

Reconstitution

Liquid concentrate to make the indicated volume.

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?