EMAIL THIS PAGE TO A FRIEND
P6938 Sigma

Brij® L23

for molecular biology

Synonym: Brij 35, C12E23, Polyoxyethylene (23) lauryl ether

Purchase

Properties

grade   for molecular biology
InChI Key   IEQAICDLOKRSRL-UHFFFAOYSA-N
foreign activity   RNase, none detected

Description

Legal Information

Brij is a registered trademark of Croda International PLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
WGK Germany 
2
RTECS 
MD0875000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P1254 Brij® L23, suitable for Stein-Moore chromatography

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?