• USA Home
  • P7026 - Protease from Aspergillus sojae

EMAIL THIS PAGE TO A FRIEND
P7026 Sigma

Protease from Aspergillus sojae

Type XIX, ≥0.3 unit/mg solid

Purchase

Description

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Protocols & Applications

Non-specific Protease Activity Assay Video

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
UK9540000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Protocols

Enzymatic Assay of Protease Using Casein As a Substrate

To standardize a procedure for the enzymatic assay of Protease using Casein as a substrate at Sigma-Aldrich St. Louis.

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?