• USA Home
  • P7042 - Carestream® Kodak® autoradiography GBX developer/replenisher

EMAIL THIS PAGE TO A FRIEND
P7042 Sigma

Carestream® Kodak® autoradiography GBX developer/replenisher

pack of 1 gal

Synonym: photo developing chemicals, photo processing chemicals

Purchase

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Compatibility

For manual processing

Quantity

Liquid concentrate to make the indicated volume.

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?