• USA Home
  • P7063 - Poly(N-methacryloyl-D-glucosamine)

EMAIL THIS PAGE TO A FRIEND
P7063 Sigma

Poly(N-methacryloyl-D-glucosamine)

Synonym: PMG

Purchase

Properties

mol wt   average mol wt 1 × 106-3 × 106 by LALLS
storage temp.   −20°C

Description

Application

Useful for the stabilization of proteins.

Other Notes

High molecular weight water-soluble polymer.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?