• USA Home
  • P7106 - Scienceware® Easyplate petri dish transport carrier

EMAIL THIS PAGE TO A FRIEND
P7106 Sigma

Scienceware® Easyplate petri dish transport carrier

size 9 in., yellow

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Dishes, Labware, Microbiology Equipment, Petri Dish Holders More...
material   polycarbonate (autoclavable)
size   9 in.
color   yellow

Description

Application

Increase workspace and efficiently transport petri dishes with the Easyplate. The Easyplate eliminates a "balancing act" of petri dishes and enables you to transport petri dishes rapidly and orderly.

Compatibility

Elastic holding devices keep the dishes in place. Write-on labels are provided for critical sample identification.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?