• USA Home
  • P7124 - Anti-Prox1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P7124 Sigma

Anti-Prox1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Prospero-related homeobox 1, Anti-Homeobox prospero-like protein PROX1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Gene Regulation, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors,
species reactivity   mouse (predicted), human
application(s)   indirect immunofluorescence: 2-4 μg/mL using human HepG2 cells
  western blot (chemiluminescent): 0.5-1 μg/mL using whole extract of human SHSY5Y cells
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt ~83 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... PROX1(5629)
mouse ... Prox1(19130)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acid residues 723-737 of human Prox1 with N-terminal lysine added, conjugated to KLH. The corresponding sequence is identical in mouse.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?