• USA Home
  • P7151 - Polygalacturonic acid lithium salt

EMAIL THIS PAGE TO A FRIEND
P7151 Sigma

Polygalacturonic acid lithium salt

from oranges, 85-90%

Synonym: Lithium polypectate, Polyanhydrogalacturonic acid

Purchase

Properties

biological source   from oranges
assay   85-90%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

P3850 Polygalacturonic acid sodium salt, from citrus fruit, ≥75% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?