• USA Home
  • P7167 - Carestream® Kodak® autoradiography GBX fixer/replenisher

EMAIL THIS PAGE TO A FRIEND
P7167 Sigma

Carestream® Kodak® autoradiography GBX fixer/replenisher

pack of 1 gal

Synonym: photo developing chemicals, photo processing chemicals

Purchase

Description

Compatibility

For manual processing

Quantity

Liquid concentrate to make the indicated volume.

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?