• USA Home
  • P7232 - Prolactin-releasing Peptide rat

EMAIL THIS PAGE TO A FRIEND
P7232 Sigma

Prolactin-releasing Peptide rat

≥97% (HPLC)

Synonym: PrRP31

  • Empirical Formula (Hill Notation) C156H242N54O43S

  • Molecular Weight 3593.99

  •  MDL number MFCD01868174

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Peptides and Proteins, Peptides for Cell Biology,
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Prlh(63850)

Description

Amino Acid Sequence

Ser-Arg-Ala-His-Gln-His-Ser-Met-Glu-Thr-Arg-Thr-Pro-Asp-Ile-Asn-Pro-Ala-Trp-Tyr-Thr-Gly-Arg-Gly-Ile-Arg-Pro-Val-Gly-Arg-Phe-NH2

Biochem/physiol Actions

Endogenous agonist for the hGR3 receptor. Releases prolactin from pituitary cultures and LH-RH from hypothalamic explants.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?