• USA Home
  • P7236 - Oxford® Benchmate® adjustable volume pipette
EMAIL THIS PAGE TO A FRIEND
P7236 Sigma
Oxford® Benchmate® adjustable volume pipette
volume 0.2-2 μL
Purchase
Description
Features and Benefits
The Oxford® Benchmate® continuously-adjustable, autoclavable pipette is a general purpose, handheld instrument used to dispense variable volumes of liquids in the laboratory environment. The entire pipette can be autoclaved (20 min at 121 °C) for decontamination and sterilization. The barrel of the BenchMate is designed to minimize dead air space for increased accuracy. A non-metallic tip ejector provides access to the bottom of most commonly used test tubes and allows for the safe disposal of used pipette tips. A lightweight, slender design reduces operator fatigue during prolonged use.
No need to recalibrate after autoclaving.
Highly resistant to chemical reagents.
Digital display allows easy volume adjustment.
Legal Information
Benchmate is a registered trademark of Oxford Worldwide LLC
Oxford is a registered trademark of Oxford Worldwide LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?