• USA Home
  • P7239 - Poly(ethyleneimine) iodide, cationic

EMAIL THIS PAGE TO A FRIEND
P7239 Sigma

Poly(ethyleneimine) iodide, cationic

≥95%, liquid

Synonym: Catimer p

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Polysaccharide More...
assay   ≥95%
form   liquid
storage temp.   2-8°C

Description

General description

Poly(ethyleneimine) iodide is a polycation.

Application

Poly(ethyleneimine) iodide has been used in a study to assess adsorption of bovine serum albumin on previously formed poly(allylamine hydrochloride)/poly(sodium 4-styrenesulphonate) multilayer.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?