• USA Home
  • P7318 - L-α-Phosphatidylcholine from frozen egg yolk

EMAIL THIS PAGE TO A FRIEND
P7318 Sigma

L-α-Phosphatidylcholine from frozen egg yolk

~99% (TLC), lyophilized powder

Purchase

Properties

assay   ~99% (TLC)
form   lyophilized powder
storage temp.   −20°C

Description

Packaging

packaged under argon

Preparation Note

Prepared by a modification of the procedure of Singleton, W.S., et al., J. Am. Oil Chem. Soc., 42, 53 (1965).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OG7565000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P3556 L-α-Phosphatidylcholine, from egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?