• USA Home
  • P7379 - 3α,17α-Dihydroxy-5β-pregnan-20-one

EMAIL THIS PAGE TO A FRIEND
P7379 Sigma

3α,17α-Dihydroxy-5β-pregnan-20-one

Synonym: 17α-Hydroxypregnanolone, 5β-Pregnane-3α,17α-diol-20-one

  • CAS Number 570-52-5

  • Empirical Formula (Hill Notation) C21H34O3

  • Molecular Weight 334.49

  •  MDL number MFCD00069487

  •  PubChem Substance ID 329820265

Purchase

Properties

InChI Key   LKQDFQLSEHWIRK-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TU4381200

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?