• USA Home
  • P7383 - Pyrophosphatase, Nucleotide from Crotalus adamanteus venom

EMAIL THIS PAGE TO A FRIEND
P7383 Sigma

Pyrophosphatase, Nucleotide from Crotalus adamanteus venom

Type II, lyophilized powder, 4-8 units/mg protein, vial of ~25 units

Synonym: Dinucleotide nucleotidohydrolase

Purchase

Properties

Related Categories 3.6.x.x Acting on acid anhydrides, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Type II
form   lyophilized powder
packaging   vial of ~25 units
foreign activity   5′-nucleotidase <0.003 unit/mg protein
  alkaline phosphatase <0.002 unit/mg protein
  phosphodiesterase 0.2 - 0.4 unit/mg protein
storage temp.   −20°C

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of β-NAD to NMN and AMP per min at pH 7.4 at 37 °C in the presence of Mg ions.

Physical form

Lyophilized powder containing approx. 35% Tris buffer salts.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?