• USA Home
  • P7557 - Carestream® Kodak® Processing chemicals

EMAIL THIS PAGE TO A FRIEND
P7557 Sigma

Carestream® Kodak® Processing chemicals

D-76 Developer

Purchase

Description

Reconstitution

Packet of powder to make the indicated volume.

Legal Information

Carestream is a registered trademark of Carestream Health, Inc.

Kodak is a registered trademark of Eastman Kodak Company

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?