• USA Home
  • P7588 - Polyacrylate-polyalcohol

EMAIL THIS PAGE TO A FRIEND
P7588 Sigma

Polyacrylate-polyalcohol

beads, ≤500 μm

Synonym: Dialyzorb

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Reagents More...
form   beads
particle size   ≤500 μm

Description

Packaging

1 kg in poly bottle

500 g in poly bottle

Application

Polyacrylate-polyalcohol is used in protein chromatography and affinity chromatography. Polyacrylate-polyalcohol has been used to study the treatment of vesicoureteric reflux (VUR).

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?