• USA Home
  • P7641 - Perchloric acid-d solution

EMAIL THIS PAGE TO A FRIEND
P7641 Sigma

Perchloric acid-d solution

68 % (w/w) in D2O

Synonym: Deuteroperchloric acid, Perchloric acid-d

Purchase

Properties

isotopic purity   ≥99 atom % D
concentration   68 % (w/w) in D2O

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1873 5.1/PG 1
RTECS 
SC7500000
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?