• USA Home
  • P7655 - Phentermine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
P7655 Sigma

Phentermine hydrochloride solution

1.0 mg/mL in methanol

Synonym: α,α-Dimethylphenethylamine hydrochloride

  • Empirical Formula (Hill Notation) C10H15N · HCl

  • Molecular Weight 185.69

Purchase

Properties

drug control   USDEA Schedule IV; Home Office Schedule 3; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?