• USA Home
  • P7657 - 1-Palmitoyl-2-(pyrene-1-yl)decanoyl-sn-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
P7657 Sigma

1-Palmitoyl-2-(pyrene-1-yl)decanoyl-sn-glycero-3-phosphocholine

≥95%

Synonym: L-α-Phosphatidylcholine, β-(pyren-1-yl)decanoyl-γ-palmitoyl

  • CAS Number 95864-17-8

  • Empirical Formula (Hill Notation) C50H76NO8P

  • Molecular Weight 850.11

  •  MDL number MFCD00063542

  •  PubChem Substance ID 24898865

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycerophospholipids, Glycerophosphonocholines, Lipids
InChI Key   HBCKORABVGGHAT-UHFFFAOYSA-N
assay   ≥95%
storage temp.   −20°C

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?