• USA Home
  • P7709 - (+)-Bis[(R)-1-phenylethyl]amine hydrochloride

EMAIL THIS PAGE TO A FRIEND
P7709 Sigma

(+)-Bis[(R)-1-phenylethyl]amine hydrochloride

≥98% (titration)

  • CAS Number 82398-30-9

  • Empirical Formula (Hill Notation) C16H19N · HCl

  • Molecular Weight 261.79

  •  MDL number MFCD00216672

  •  PubChem Substance ID 24898869

Purchase

Properties

Related Categories Amine Salts, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   ZBQCLJZOKDRAOW-DTPOWOMPSA-N
assay   ≥98% (titration)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?