• USA Home
  • P7735 - Eppendorf® Research® Pro electronic pipette, single channel
EMAIL THIS PAGE TO A FRIEND
P7735 Sigma
Eppendorf® Research® Pro electronic pipette, single channel
without charging adapter, volume range 20-300
Purchase
Properties
Related Categories Eppendorf, Eppendorf Pipettors, Eppendorf Pipettors & Tips, Labware, Liquid Handling Equipment,
mfr. no.   Eppendorf®, 022461371 (USA)
  Eppendorf®, 4860 000.089 (European)
  Eppendorf®, 4860 000.089
volume range   20-300
Description
General description
This pipette provides a wide variety of programmable application parameters, while still remaining simple to use and easy to handle. The styling of the Eppendorf Research pro pipette facilitates fast and accurate performance, with the expected precision and reliability of Eppendorf
• Easy-to-read display screen is set at a 45° angle for easy reference during use
• Motorized piston operation increases reproducibility
• All buttons and controls are large and easily accessible for both right- and left-handed users
• Volume and speed options are controlled with a rocker switch, requiring only a minimal amount of force avoids potential wrist and hand strain
• Separate tip ejection lever-can be adjusted to the most comfortable position for right- and left-handed users
• Fully charged battery performs over 2,000 pipetting cycles-reduces downtime
• Automatic shut-off function after 10 minutes of non-use
• Color coded actuate button corresponds with common pipette tip volume sizes
• User-calibratable-ideal for use with solutions of different viscosity, density, surface tension, or vapor pressure
• Lower half is removable and autoclavable at 121°C - for simplified decontamination
• Quick recharge NiMH battery performs up to 200 pipetting strokes after recharging only 30 minutes
• For each program, 10 individual aspirating and dispensing speeds can be programmed.
Legal Information
Eppendorf Research is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?