• USA Home
  • P7752 - 6-Phosphogluconic acid tri(cyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
P7752 Sigma

6-Phosphogluconic acid tri(cyclohexylammonium) salt

Grade V

Synonym: 6-Phosphogluconic acid tris(cyclohexylamine) salt, D-Gluconate 6-phosphate tris(cyclohexylamine) salt

  • CAS Number 108347-81-5

  • Empirical Formula (Hill Notation) C6H13O10P · 3C6H13N

  • Molecular Weight 573.66

  •  MDL number MFCD00069563

  •  PubChem Substance ID 24898871

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
InChI Key   SLRCCWJSBJZJBV-ZMYVRHLSSA-N
type   Grade V
storage temp.   −20°C

Description

Preparation Note

Prepared by a modification of the procedure of Seegmiller, J.E., et al., J. Biol. Chem., 192, 175 (1951).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Pentose Phosphate Pathway

While glucose metabolism by glycolysis occurs where energy is needed quickly, e.g. in brain and muscle cells, a second pathway for glucose metabolism, called pentose phosphate pathway, operates in ti...
BioFiles 2007, 2.6, 25.
Keywords: Catalysis, Glycolysis, Metabolism, Oxidations, Pentose phosphate pathway

Related Products

replaced by

Product #

Description

Add to Cart

P7877 6-Phosphogluconic acid trisodium salt, ≥97% (enzymatic)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?