• USA Home
  • P7763 - L-α-Phosphatidylcholine from bovine liver

EMAIL THIS PAGE TO A FRIEND
P7763 Sigma

L-α-Phosphatidylcholine from bovine liver

Type III-L, 10 mg/mL in chloroform: methanol (3:1), ~99%

Purchase

Properties

type   Type III-L
assay   ~99%
concentration   10 mg/mL in chloroform: methanol (3:1)
storage temp.   −20°C

Description

Preparation Note

Prepared chromatographically

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2929 6.1/PG 2
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?