• USA Home
  • P7796 - Blotting paper, pure cellulose
EMAIL THIS PAGE TO A FRIEND
P7796 Sigma
Blotting paper, pure cellulose
Extra Thick, size 7 cm × 10 cm
Purchase
Description
General description
Blotting paper is used for Northern, Southern and Western blots. This is a clean and fast alternative to paper towels for Southern and Northern capillary transfer. Ten sheets of the extra thick blotting paper will produce complete capillary transfer in two hours. Ideal for Western blotting to sandwich the gel and the membrane.
Price and Availability
Antibody Explorer
Explore with Confidence
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?