• USA Home
  • P7832 - N-26 Sequencing Primer

EMAIL THIS PAGE TO A FRIEND
P7832 Sigma

N-26 Sequencing Primer

Purchase

Properties

shipped in   dry ice
storage temp.   −20°C

Description

Application

Sequencing primers are used for double-stranded DNA sequencing of inserts in bacterial FLAG expression vectors.

The N-26 primer is used for sequencing DNA at the 5′ end of the ORF in E. coli FLAG® expression vectors. This corresponds to the amino-terminal fusion junction of FLAG fusion proteins. It can also be used to sequence the amino-terminus of carboxy-terminal FLAG fusion proteins. Complementary sequence is immediately 5′ to the tac promoter region.

Other Notes

(5′-CAT CAT AAC GGT TCT GGC AAA TAT TC-3′)

Legal Information

FLAG is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?