EMAIL THIS PAGE TO A FRIEND
P7863 Sigma

Poly(A)+ RNA human

from human normal adult (Heart)

Purchase

Properties

Related Categories Cell Biology, Expression/Translation, Gene Expression & Analysis, General Stem Cell Biology, Molecular Biology,
biological source   from human normal adult (Heart)
shipped in   dry ice
storage temp.   −70°C

Description

Other Notes

Human mRNA is isolated from total RNA by affinity chromatography using oligo(dT) columns. Following isolation, the integrity of the mRNA is verified on a denaturing agarose gel. All mRNAs show very high efficiencies in reverse transcription.

Packaging

Each vial contains 5 μg of poly(A)+ RNA in DEPC treated water

Legal Information

Manufactured by BioChain, Inc.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?