EMAIL THIS PAGE TO A FRIEND
P7967 Sigma

[Ala92]-Peptide 6

Synonym: [Ala92]-p16 fragment 84-103

Purchase

Description

Amino Acid Sequence

Asp-Ala-Ala-Arg-Glu-Gly-Phe-Leu-Ala-Thr-Leu-Val-Val-Leu-His-Arg-Ala-Gly-Ala-Arg

Biochem/physiol Actions

Modified fragment from the third ankyrin-like repeat of p16 that binds to cdk4 and cdk6 and inhibits phosphorylation of the retinoblastoma gene product pRb mediated by cdk4-cyclin D.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?