• USA Home
  • P8023 - Phospholipase D from Streptomyces chromofuscus

EMAIL THIS PAGE TO A FRIEND
P8023 Sigma

Phospholipase D from Streptomyces chromofuscus

lyophilized powder, ≥2,000 units/mg solid

Synonym: Lecithinase D, Phosphatidylcholine phosphatidohydrolase

Purchase

Description

Biochem/physiol Actions

Phospholipase D is active on sphingomyelins and lysophospholipids. Phospholipase D hydrolyzes the phosphate bonds of phospholipids and sphingomyelin to give the corresponding phosphatidic acid.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

One unit will liberate 1.0 μM of choline from L-α-phosphatidylcholine (egg yolk) per hr at pH 8.0 at 30°C.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P0065 Phospholipase D from Streptomyces chromofuscus, ≥50,000 units/mL, buffered aqueous glycerol solution

inhibitor

Product #

Description

Add to Cart

D6518 Diethylenetriaminepentaacetic acid, ≥99% (titration)
P9375 1,10-Phenanthroline monohydrate, reagent grade
E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
ED2SS Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?