• USA Home
  • P8036 - Protein A–Acrylic beads from Staphylococcus aureus

EMAIL THIS PAGE TO A FRIEND
P8036 Sigma

Protein A–Acrylic beads from Staphylococcus aureus

lyophilized powder, 150 μm

Purchase

Properties

form   lyophilized powder
matrix   acrylic beads
matrix activation   oxirane
matrix attachment   amino
matrix spacer   3 atoms
particle size   150 μm
capacity   ~5 mg/mL binding capacity (human IgG)
storage temp.   −20°C

Description

Quantity

Swelling: 1 g swells to approx. 4 ml

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?