• USA Home
  • P8040 - D-Phenylalanine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
P8040 Sigma

D-Phenylalanine methyl ester hydrochloride

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
InChI Key   SWVMLNPDTIFDDY-SBSPUUFOSA-N
mp   159-163 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

525472 D-Phenylalanine methyl ester hydrochloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?