• USA Home
  • P8090 - Anti-Protein Kinase Cη antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P8090 Sigma

Anti-Protein Kinase Cη antibody produced in rabbit

whole antiserum

Synonym: Anti-PKC η

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, PR-PR,
species reactivity   human
application(s)   microarray: suitable
  western blot: 1:2,000 using whole cell lysate of mouse NIH3T3 fibroblasts overexpressing human PKCη
clone   polyclonal
antibody form   whole antiserum
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... PRKCH(5583)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the C-terminal variable (V5) region of PKC η (amino acids 670-683).

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?