• USA Home
  • P8091 - Precast Mini-Polyacrylamide Gels 10-20% Tricine gradient gel

EMAIL THIS PAGE TO A FRIEND
P8091 Sigma

Precast Mini-Polyacrylamide Gels 10-20% Tricine gradient gel

Purchase

Properties

cassette H × W × thickness   10 cm × 10 cm × 5 mm
gel H × W × thickness   8.8 cm × 8.1 cm × 1 mm
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?