EMAIL THIS PAGE TO A FRIEND
P8092 Sigma

Cyclin A primer set

Purchase

Properties

storage temp.   −20°C

Description

Application

Amplifies human and hamster.

Synthetic oligonucleotide primers for the amplification of specific genes by PCR. Each set includes a 5′-sense and a 3′-antisense oligonucleotide in separate tubes ready for use in PCR. The primer sets can also be used for the detection of specific gene expression by RT-PCR.

Packaging

Sets include enough of the sense and anti-sense primers for 50 reactions. Information on optimized working conditions is provided.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?