• USA Home
  • P8107 - Anti-P2X4 Purinergic Receptor antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P8107 Sigma

Anti-P2X4 Purinergic Receptor antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurotransmission, Antibodies to Ion Channels,
species reactivity   rat
application(s)   immunohistochemistry: suitable using rat brain sections
  western blot (chemiluminescent): 1:200-1:300 using rat brain lysate
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... P2rx4(29659)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

peptide (C)KKYK YVEDY EQGLS GEMNQ corresponding to amino acids 370-388 of rat P2X4

Physical form

After reconstitution, antibody is in phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?